Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153614.3 | NP_705842.2 | 2 | 855 | ||
XM_005273984.3 | XP_005274041.1 | 2 | 382 | |||
XM_011545004.2 | XP_011543306.1 | 4 | 1106 | |||
XM_011545005.2 | XP_011543307.1 | 4 | 1108 | |||
XM_011545007.2 | XP_011543309.1 | 1 | 343 | |||
XM_011545009.2 | XP_011543311.1 | 3 | 649 | |||
XM_011545013.2 | XP_011543315.1 | 4 | 1112 | |||
XM_011545014.2 | XP_011543316.1 | 4 | 1112 | |||
XM_017017675.1 | XP_016873164.1 | 2 | 88 | |||
XM_017017676.1 | XP_016873165.1 | 1 | 344 | |||
XM_017017677.1 | XP_016873166.1 | 1 | 344 | |||
XM_017017678.1 | XP_016873167.1 | 1 | 344 | |||
XM_017017679.1 | XP_016873168.1 | 4 | 1112 | |||
GenBank mRNA | AK128589.1 | - | 2 | 148 | ||
AY138810.2 | - | 2 | 855 | |||
AY325766.1 | - | 2 | 148 |