Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIX1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153713.2 | NP_714924.1 | 4 | 713 | ||
GenBank mRNA | AK128733.1 | - | 5 | 554 | ||
BC035727.1 | - | 4 | 695 | 5723677 | ||
CR627381.1 | - | 3 | 351 |