Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIX1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153713.2 | NP_714924.1 | 5, 6 | 874 | ||
XM_017000316.1 | XP_016855805.1 | 4, 5 | 797 | |||
GenBank mRNA | AK123675.1 | - | 2, 3 | 126 | ||
AK128733.1 | - | 6, 7 | 715 | |||
BC035727.1 | - | 5, 6 | 856 | 5723677 | ||
CR627381.1 | - | 4, 5 | 512 |