Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321335.1 | NP_001308264.1 | 4 | 849 | ||
NM_170746.3 | NP_734467.1 | 4 | 869 | |||
GenBank mRNA | AF536829.1 | - | 4 | 765 | ||
BC021122.1 | - | 4 | 583 | 4766360 | ||
BC111087.1 | - | 3 | 703 | |||
BG714263.1 | - | 4 | 564 |