Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM163A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173509.2 | NP_775780.1 | 5 | 1304 | ||
XM_005244913.2 | XP_005244970.1 | 6 | 1741 | |||
XM_006711174.2 | XP_006711237.1 | 6 | 1452 | |||
XM_006711175.2 | XP_006711238.1 | 7 | 1564 | |||
XM_006711176.2 | XP_006711239.1 | 5 | 1430 | |||
XM_006711177.1 | XP_006711240.1 | 6 | 1284 | |||
XM_011509217.1 | XP_011507519.1 | 8 | 1586 | |||
XM_011509221.1 | XP_011507523.1 | 7 | 1392 | |||
XM_011509226.1 | XP_011507528.1 | 5 | 1226 | |||
XM_011509228.1 | XP_011507530.1 | 5 | 1360 | |||
XM_017000386.1 | XP_016855875.1 | 8 | 1735 | |||
XM_017000387.1 | XP_016855876.1 | 9 | 1857 | |||
XM_017000388.1 | XP_016855877.1 | 7 | 1598 | |||
XM_017000389.1 | XP_016855878.1 | 9 | 1869 | |||
XM_017000390.1 | XP_016855879.1 | 7 | 1589 | |||
XM_017000391.1 | XP_016855880.1 | 7 | 1713 | |||
XM_017000392.1 | XP_016855881.1 | 6 | 1238 | |||
XM_017000393.1 | XP_016855882.1 | 5 | 1097 | |||
GenBank mRNA | AK310711.1 | - | 5 | 1360 | ||
AY902195.1 | - | 5 | 1304 | |||
BC009382.2 | - | 5 | 1304 | 4124490 |