Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286159.1 | NP_001273088.1 | 7 | 1147 | ||
NM_173556.4 | NP_775827.2 | 7 | 1147 | |||
XM_011544839.2 | XP_011543141.1 | 7 | 733 | |||
XM_011544840.2 | XP_011543142.1 | 7 | 805 | |||
XM_011544841.1 | XP_011543143.1 | 7 | 695 | |||
XM_011544842.1 | XP_011543144.1 | 7 | 1147 | |||
XM_011544843.1 | XP_011543145.1 | 6 | 979 | |||
XM_011544844.2 | XP_011543146.1 | 5 | 859 | |||
XM_017017354.1 | XP_016872843.1 | 7 | 1147 | |||
XM_017017355.1 | XP_016872844.1 | 6 | 979 | |||
GenBank mRNA | AK093405.1 | - | 5 | 527 | ||
AK124113.1 | - | 7 | 1148 | |||
AK314036.1 | - | 7 | 851 | |||
AY251167.1 | - | 4 | 353 | |||
BC040280.1 | - | 7 | 825 | 5166625 |