Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278736.1 | NP_001265665.1 | 2 | 220 | ||
NM_002985.2 | NP_002976.2 | 2 | 220 | |||
GenBank mRNA | AF043341.1 | - | 2 | 155 | ||
AF266753.1 | - | 2 | 152 | |||
AK312212.1 | - | 2 | 207 | |||
BC008600.1 | - | 2 | 176 | 4185200 | ||
CR980938.1 | - | 2 | 138 | |||
DQ230537.1 | - | 2 | 152 | |||
M21121.1 | - | 2 | 178 |