Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080407.2 | NP_001073876.2 | 19 | 2181 | ||
XM_011542566.2 | XP_011540868.1 | 18 | 2251 | |||
XM_011542567.2 | XP_011540869.1 | 18 | 2251 | |||
XM_011542568.2 | XP_011540870.1 | 18 | 2215 | |||
XM_011542569.2 | XP_011540871.1 | 14 | 1840 | |||
XM_011542570.2 | XP_011540872.1 | 14 | 1804 | |||
XM_011542572.1 | XP_011540874.1 | 14 | 1399 | |||
XM_011542573.1 | XP_011540875.1 | 12 | 1335 | |||
XM_011542574.2 | XP_011540876.1 | 12 | 1458 | |||
XM_017017155.1 | XP_016872644.1 | 19 | 3079 | |||
XM_017017156.1 | XP_016872645.1 | 15 | 2704 | |||
GenBank mRNA | AK074712.1 | - | 10 | 887 | ||
AK124514.1 | - | 5 | 3100 | |||
BC131702.1 | - | 9 | 798 |