Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLB1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080407.2 | NP_001073876.2 | 20 | 2911 | ||
XM_011542566.2 | XP_011540868.1 | 19 | 2981 | |||
XM_011542568.2 | XP_011540870.1 | 19 | 2945 | |||
XM_011542569.2 | XP_011540871.1 | 15 | 2570 | |||
XM_011542570.2 | XP_011540872.1 | 15 | 2534 | |||
XM_011542572.1 | XP_011540874.1 | 15 | 2129 | |||
XM_011542573.1 | XP_011540875.1 | 13 | 2065 | |||
XM_011542574.2 | XP_011540876.1 | 13 | 2188 | |||
XM_017017155.1 | XP_016872644.1 | 20 | 3809 | |||
XM_017017156.1 | XP_016872645.1 | 16 | 3434 | |||
GenBank mRNA | AK074712.1 | - | 11 | 1618 | ||
AK124514.1 | - | 6 | 3830 |