Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173588.3 | NP_775859.3 | 20 | 3336 | ||
XM_011520030.2 | XP_011518332.1 | 18 | 6240 | |||
XM_011520031.2 | XP_011518333.1 | 17 | 5937 | |||
XM_017017607.1 | XP_016873096.1 | 19 | 6131 | |||
XM_017017608.1 | XP_016873097.1 | 19 | 4154 | |||
GenBank mRNA | AK095113.1 | - | 18 | 2944 | ||
BC115397.1 | - | 19 | 3486 | 40068579 |