Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130957.1 | NP_001124429.1 | 2 | 147 | ||
NM_173807.4 | NP_776168.1 | 2 | 147 | |||
XM_011544142.2 | XP_011542444.1 | 3 | 421 | |||
XM_011544149.2 | XP_011542451.1 | 2 | 83 | |||
XM_011544151.2 | XP_011542453.1 | 2 | 114 | |||
XM_017000945.1 | XP_016856434.1 | 2 | 200 | |||
XM_017000948.1 | XP_016856437.1 | 2 | 92 | |||
GenBank mRNA | AK027205.1 | - | 3 | 337 | ||
AK302609.1 | - | 2 | 100 | |||
BC032859.1 | - | 2 | 132 | 5269307 | ||
DB462258.1 | - | 2 | 202 |