Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130957.1 | NP_001124429.1 | 8 | 541 | ||
NM_001242340.1 | NP_001229269.1 | 5 | 389 | |||
NM_173807.4 | NP_776168.1 | 8 | 541 | |||
XM_011544138.2 | XP_011542440.1 | 8 | 592 | |||
XM_011544139.2 | XP_011542441.1 | 7 | 487 | |||
XM_011544142.2 | XP_011542444.1 | 9 | 815 | |||
XM_011544144.2 | XP_011542446.1 | 7 | 518 | |||
XM_011544145.2 | XP_011542447.1 | 6 | 412 | |||
XM_011544149.2 | XP_011542451.1 | 6 | 374 | |||
XM_017000943.1 | XP_016856432.1 | 8 | 593 | |||
XM_017000946.1 | XP_016856435.1 | 7 | 692 | |||
XM_017000947.1 | XP_016856436.1 | 8 | 593 | |||
XM_017000948.1 | XP_016856437.1 | 9 | 537 | |||
XM_017000949.1 | XP_016856438.1 | 7 | 742 | |||
XM_017000950.1 | XP_016856439.1 | 6 | 388 | |||
XM_017000951.1 | XP_016856440.1 | 8 | 593 | |||
XM_017000952.1 | XP_016856441.1 | 8 | 593 | |||
XM_017000953.1 | XP_016856442.1 | 8 | 594 | |||
GenBank mRNA | AK302609.1 | - | 8 | 494 | ||
AK303059.1 | - | 5 | 349 | |||
BC032859.1 | - | 8 | 526 | 5269307 |