Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLRAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304263.1 | NP_001291192.1 | 5 | 2143 | ||
NM_001304264.1 | NP_001291193.1 | 6 | 2259 | |||
NM_174902.3 | NP_777562.1 | 6 | 2290 | |||
GenBank mRNA | AK075546.1 | - | 6 | 2209 | ||
BC042754.1 | - | 6 | 2193 | 4828259 |