Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175058.4 | NP_778228.3 | 4 | 283 | ||
XM_017017240.1 | XP_016872729.1 | 4 | 290 | |||
XM_017017241.1 | XP_016872730.1 | 4 | 290 | |||
XM_017017242.1 | XP_016872731.1 | 4 | 290 | |||
XM_017017243.1 | XP_016872732.1 | 4 | 290 | |||
GenBank mRNA | BC071599.1 | - | 4 | 279 | 30343461 | |
CN293588.1 | - | 4 | 283 |