Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STPG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199012.1 | NP_001185941.1 | 6 | 771 | ||
NM_001199013.1 | NP_001185942.1 | 6 | 683 | |||
NM_001199014.1 | NP_001185943.1 | 7 | 827 | |||
NM_178122.4 | NP_835223.1 | 5 | 564 | |||
XM_006711025.2 | XP_006711088.1 | 7 | 789 | |||
XM_011542403.2 | XP_011540705.1 | 5 | 1088 | |||
XM_011542404.1 | XP_011540706.1 | 7 | 902 | |||
XM_017002768.1 | XP_016858257.1 | 7 | 898 | |||
XM_017002769.1 | XP_016858258.1 | 8 | 1046 | |||
XM_017002770.1 | XP_016858259.1 | 9 | 1161 | |||
GenBank mRNA | AK025957.1 | - | 4 | 494 | ||
BC017650.1 | - | 2 | 90 | |||
BC035061.1 | - | 7 | 800 | 5262741 | ||
BC047705.1 | - | 4 | 469 | 5764388 | ||
BC063891.1 | - | 6 | 715 | 30390664 | ||
BM922375.1 | - | 6 | 646 |