Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RILPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319243.1 | NP_001306172.1 | 5 | 1201 | ||
NM_001319244.1 | NP_001306173.1 | 5 | 750 | |||
NM_001319302.1 | NP_001306231.1 | 6 | 1700 | |||
NM_178314.4 | NP_847884.2 | 5 | 1201 | |||
XM_006719372.2 | XP_006719435.1 | 5 | 1195 | |||
XM_006719373.3 | XP_006719436.1 | 4 | 548 | |||
XM_011538282.2 | XP_011536584.1 | 5 | 1236 | |||
XM_011538284.2 | XP_011536586.1 | 4 | 767 | |||
XM_011538285.2 | XP_011536587.1 | 4 | 528 | |||
GenBank mRNA | AK096697.1 | - | 6 | 1043 | ||
BC080626.1 | - | 5 | 750 | 6339887 | ||
BC089444.1 | - | 5 | 1003 | 30559974 |