Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FERMT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031471.5 | NP_113659.3 | 11 | 1369 | ||
NM_178443.2 | NP_848537.1 | 11 | 1381 | |||
XM_011545294.2 | XP_011543596.1 | 11 | 1393 | |||
XM_011545295.2 | XP_011543597.1 | 10 | 1226 | |||
XM_017018398.1 | XP_016873887.1 | 11 | 1377 | |||
XM_017018399.1 | XP_016873888.1 | 10 | 1229 | |||
GenBank mRNA | AK093719.1 | - | 11 | 1314 | ||
AY093951.1 | - | 11 | 1315 | |||
AY093952.1 | - | 11 | 1327 | |||
BC004347.1 | - | 11 | 1313 | 3634032 | ||
BC013366.2 | - | 11 | 1313 | 3677274 | ||
BC015584.2 | - | 11 | 1308 | 4637808 |