Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FERMT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031471.5 | NP_113659.3 | 15 | 2025 | ||
NM_178443.2 | NP_848537.1 | 15 | 2037 | |||
XM_011545294.2 | XP_011543596.1 | 15 | 2049 | |||
XM_011545295.2 | XP_011543597.1 | 14 | 1882 | |||
XM_017018398.1 | XP_016873887.1 | 15 | 2033 | |||
XM_017018399.1 | XP_016873888.1 | 14 | 1885 | |||
GenBank mRNA | AK093719.1 | - | 15 | 1970 | ||
AY093951.1 | - | 15 | 1971 | |||
AY093952.1 | - | 15 | 1983 | |||
BC004347.1 | - | 15 | 1969 | 3634032 | ||
BC013366.2 | - | 15 | 1969 | 3677274 | ||
BC015584.2 | - | 15 | 1964 | 4637808 | ||
BG491422.1 | - | 2 | 207 |