Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAYN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258390.1 | NP_001245319.1 | 3 | 583 | ||
NM_001258391.1 | NP_001245320.1 | 2 | 559 | |||
NM_001318799.1 | NP_001305728.1 | 2 | 193 | |||
NM_178834.4 | NP_849156.1 | 2 | 559 | |||
XM_006718769.3 | XP_006718832.1 | 2 | 172 | |||
XM_006718770.3 | XP_006718833.1 | 2 | 387 | |||
GenBank mRNA | AK055539.1 | - | 2 | 560 | ||
AK055654.1 | - | 2 | 347 | |||
AK293317.1 | - | 2 | 390 | |||
AK296231.1 | - | 2 | 347 | |||
AY358554.1 | - | 3 | 408 | |||
BC025407.1 | - | 2 | 352 | 4820414 | ||
DB006476.1 | - | 2 | 413 | |||
DC398135.1 | - | 2 | 193 |