Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181706.4 | NP_859057.4 | 5 | 705 | ||
XM_017017216.1 | XP_016872705.1 | 5 | 1478 | |||
GenBank mRNA | AL050199.1 | - | 1 | 92 | ||
AL833128.1 | - | 5 | 695 | |||
BC044931.2 | - | 5 | 657 | |||
BC063804.1 | - | 5 | 648 | 6474523 |