Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025598.1 | NP_001020769.1 | 10 | 1440 | ||
NM_001287600.1 | NP_001274529.1 | 9 | 1295 | |||
NM_001287602.1 | NP_001274531.1 | 6 | 909 | |||
NM_181720.2 | NP_859071.2 | 10 | 1440 | |||
XM_005245070.2 | XP_005245127.1 | 10 | 1440 | |||
XM_005245073.3 | XP_005245130.1 | 8 | 1193 | |||
XM_011509391.2 | XP_011507693.1 | 9 | 1271 | |||
GenBank mRNA | AK126116.1 | - | 10 | 1435 | ||
AK126163.1 | - | 6 | 909 | |||
AK160366.1 | - | 10 | 1415 | |||
AL832852.1 | - | 8 | 875 | |||
BC043387.1 | - | 9 | 1128 | |||
BC053688.1 | - | 10 | 1136 | 5213184 | ||
BX537846.1 | - | 8 | 1876 |