Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276504.1 | NP_001263433.1 | 2 | 184 | ||
NM_001276506.1 | NP_001263435.1 | 3 | 301 | |||
NM_003002.3 | NP_002993.1 | 3 | 301 | |||
NR_077060.1 | 3 | 301 | ||||
GenBank mRNA | AB006202.1 | - | 3 | 228 | ||
AK075360.1 | - | 3 | 227 | |||
BC005263.1 | - | 3 | 245 | 3930691 | ||
BC009574.1 | - | 3 | 226 | 4063808 | ||
BC012603.1 | - | 3 | 230 | 4108104 | ||
BC015188.1 | - | 3 | 252 | 4279214 | ||
BC015992.1 | - | 3 | 248 | 4292983 | ||
BC022350.1 | - | 3 | 240 | 4275516 | ||
BC071755.1 | - | 3 | 244 | 4774525 | ||
BC071756.1 | - | 3 | 226 | 6450992 | ||
BG492859.1 | - | 3 | 228 | |||
BQ932668.1 | - | 3 | 309 | |||
BT007238.1 | - | 3 | 217 | |||
BX419391.2 | - | 3 | 278 | |||
CR456932.1 | - | 3 | 217 | |||
KU178325.1 | - | 3 | 217 |