Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256947.1 | NP_001243876.1 | 3 | 419 | ||
NM_001282670.1 | NP_001269599.1 | 8 | 3401 | |||
NM_001282671.1 | NP_001269600.1 | 5 | 686 | |||
NM_001282672.1 | NP_001269601.1 | 4 | 555 | |||
NM_182533.2 | NP_872339.2 | 4 | 555 | |||
NR_046427.1 | 4 | 748 | ||||
XM_006710419.3 | XP_006710482.1 | 6 | 1271 | |||
XM_006710421.3 | XP_006710484.1 | 5 | 661 | |||
XM_011540914.2 | XP_011539216.1 | 10 | 1222 | |||
XM_011540921.2 | XP_011539223.1 | 7 | 1071 | |||
XM_011540922.1 | XP_011539224.1 | 5 | 685 | |||
XM_017000553.1 | XP_016856042.1 | 8 | 1069 | |||
XM_017000557.1 | XP_016856046.1 | 4 | 555 | |||
GenBank mRNA | AF469129.1 | - | 5 | 652 | ||
AK054818.1 | - | 1 | 2025 | |||
AK055593.1 | - | 4 | 555 | |||
AK125445.1 | - | 1 | 2872 | |||
AK126870.1 | - | 8 | 3400 | |||
AK128507.1 | - | 1 | 3859 | |||
AL545931.3 | - | 4 | 711 | |||
BC066360.1 | - | 4 | 552 | |||
BC103992.2 | - | 4 | 555 | 40022365 | ||
BU542153.1 | - | 3 | 380 | |||
CN397640.1 | - | 3 | 398 | |||
DA596011.1 | - | 4 | 555 |