Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182581.3 | NP_872387.2 | 3 | 795 | ||
XM_005245103.3 | XP_005245160.1 | 2 | 789 | |||
XM_011509443.2 | XP_011507745.1 | 3 | 960 | |||
XM_011509444.1 | XP_011507746.1 | 3 | 807 | |||
GenBank mRNA | AY248900.1 | - | 3 | 775 | ||
BC032957.1 | - | 3 | 802 | 4827863 |