Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318818.1 | NP_001305747.1 | 7 | 3758 | ||
NM_001318820.1 | NP_001305749.1 | 6 | 3816 | |||
NM_001318821.1 | NP_001305750.1 | 7 | 3924 | |||
NM_001318822.1 | NP_001305751.1 | 5 | 3739 | |||
NM_001318823.1 | NP_001305752.1 | 6 | 3847 | |||
NM_001318824.1 | NP_001305753.1 | 7 | 3727 | |||
NM_182612.3 | NP_872418.1 | 8 | 3835 | |||
NR_134867.1 | 7 | 3866 | ||||
NR_134868.1 | 8 | 3929 | ||||
GenBank mRNA | AK091602.1 | - | 9 | 2058 | ||
AK092816.1 | - | 9 | 1673 | |||
AK094678.1 | - | 2 | 1895 | |||
AK128653.1 | - | 6 | 3804 |