Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182833.1 | NP_878253.1 | 12 | 1361 | ||
XM_011544834.1 | XP_011543136.1 | 13 | 1587 | |||
GenBank mRNA | AY313782.1 | - | 12 | 1361 | ||
AY326450.1 | - | 12 | 1361 |