Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077262.1 | NP_001070730.1 | 8 | 829 | ||
NM_145345.2 | NP_663320.2 | 12 | 1309 | |||
NM_183008.2 | NP_892120.2 | 13 | 1314 | |||
GenBank mRNA | AF370425.1 | - | 2 | 1102 | ||
AF520989.1 | - | 11 | 1018 | |||
AF521017.1 | - | 11 | 1019 | |||
AF521018.1 | - | 11 | 1006 | |||
AK074358.1 | - | 7 | 1823 | |||
AK090792.1 | - | 7 | 1788 | |||
AK093258.1 | - | 13 | 1258 | |||
AK094198.1 | - | 8 | 761 | |||
AK097368.1 | - | 12 | 1407 | |||
AY007163.1 | - | 9 | 773 | |||
BC038106.1 | - | 11 | 2107 | 5269682 | ||
BX647774.1 | - | 11 | 2103 | |||
BX648412.1 | - | 10 | 2004 | |||
CR749413.1 | - | 11 | 2137 |