Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTY2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194285.2 | NP_919261.2 | 3 | 551 | ||
XM_011519919.2 | XP_011518221.1 | 3 | 409 | |||
GenBank mRNA | AK096760.1 | - | 3 | 571 | ||
AK304078.1 | - | 3 | 467 | |||
BC056261.1 | - | 3 | 426 | 6616686 | ||
BX641102.1 | - | 3 | 452 | |||
BX647798.1 | - | 3 | 551 | |||
BX648114.1 | - | 1 | 130 | |||
BX648478.1 | - | 3 | 473 |