Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256404.1 | NP_001243333.1 | 4 | 1131 | ||
NM_198459.3 | NP_940861.3 | 2 | 762 | |||
GenBank mRNA | AK094418.1 | - | 1 | 810 | ||
AK308302.1 | - | 2 | 783 | |||
BX647950.1 | - | 2 | 763 | |||
CR749576.1 | - | 4 | 1055 |