Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320257.1 | NP_001307186.1 | 6 | 1186 | ||
NM_198460.2 | NP_940862.2 | 8 | 1527 | |||
XM_011540835.2 | XP_011539137.1 | 8 | 1356 | |||
GenBank mRNA | AK131356.1 | - | 8 | 1511 | ||
AK290303.1 | - | 8 | 1347 | |||
AK299408.1 | - | 7 | 1162 | |||
AK299466.1 | - | 6 | 1003 | |||
AK303987.1 | - | 8 | 1271 | |||
AK315834.1 | - | 8 | 1345 | |||
BC131713.1 | - | 8 | 1305 | 40122722 | ||
BX537949.1 | - | 8 | 1346 | |||
BX647907.1 | - | 8 | 1344 |