Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320257.1 | NP_001307186.1 | 9 | 1802 | ||
NM_198460.2 | NP_940862.2 | 11 | 2143 | |||
XM_011540835.2 | XP_011539137.1 | 11 | 1972 | |||
GenBank mRNA | AK131329.1 | - | 6 | 1025 | ||
AK131356.1 | - | 11 | 2127 | |||
AK290303.1 | - | 11 | 1963 | |||
AK299408.1 | - | 10 | 1778 | |||
AK299466.1 | - | 9 | 1619 | |||
AK303987.1 | - | 11 | 1887 | |||
AK315834.1 | - | 11 | 1961 | |||
BC131713.1 | - | 11 | 1921 | 40122722 | ||
BX537949.1 | - | 11 | 1962 | |||
BX647907.1 | - | 11 | 1960 |