Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198474.3 | NP_940876.2 | 3 | 2617 | ||
XM_005252878.4 | XP_005252935.1 | 4 | 2394 | |||
GenBank mRNA | AY358591.1 | - | 3 | 2219 | ||
BC092488.1 | - | 3 | 2462 | 30531479 | ||
BX647908.1 | - | 3 | 2505 |