Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198485.3 | NP_940887.1 | 2 | 259 | ||
XM_005247382.4 | XP_005247439.1 | 2 | 3503 | |||
XM_005247388.3 | XP_005247445.1 | 2 | 3515 | |||
XM_011512732.2 | XP_011511034.2 | 2 | 2329 | |||
XM_011512734.2 | XP_011511036.1 | 2 | 3509 | |||
XM_011512735.2 | XP_011511037.1 | 2 | 3504 | |||
XM_011512736.2 | XP_011511038.1 | 2 | 3515 | |||
XM_017006257.1 | XP_016861746.1 | 9 | 1491 | |||
XM_017006258.1 | XP_016861747.1 | 6 | 919 | |||
XM_017006259.1 | XP_016861748.1 | 8 | 2102 | |||
XM_017006260.1 | XP_016861749.1 | 8 | 1298 | |||
XM_017006261.1 | XP_016861750.1 | 6 | 1107 | |||
XM_017006262.1 | XP_016861751.1 | 1 | 103 | |||
GenBank mRNA | AK123232.1 | - | 7 | 1318 | ||
AK125682.1 | - | 2 | 260 | |||
BC101550.1 | - | 2 | 160 | 8069056 | ||
BC101552.1 | - | 2 | 160 | 8069058 |