Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198493.2 | NP_940895.1 | 2 | 342 | ||
XM_011509466.2 | XP_011507768.1 | 3 | 789 | |||
XM_011509467.1 | XP_011507769.1 | 4 | 1313 | |||
XM_011509468.1 | XP_011507770.1 | 3 | 1281 | |||
XM_011509470.1 | XP_011507772.1 | 2 | 997 | |||
XM_011509471.2 | XP_011507773.1 | 3 | 1981 | |||
XM_017001123.1 | XP_016856612.1 | 4 | 651 | |||
XM_017001124.1 | XP_016856613.1 | 3 | 1981 | |||
GenBank mRNA | AK127170.1 | - | 2 | 342 | ||
BC094694.1 | - | 2 | 312 | |||
BC126353.1 | - | 2 | 342 | 8992069 | ||
BC126355.1 | - | 2 | 342 | 8992071 |