Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272046.1 | NP_001258975.1 | 4 | 540 | ||
NM_001320804.1 | NP_001307733.1 | 4 | 482 | |||
XM_011539754.2 | XP_011538056.1 | 4 | 539 | |||
XM_011539757.2 | XP_011538059.1 | 4 | 812 | |||
XM_017016177.1 | XP_016871666.1 | 4 | 292 | |||
XM_017016179.1 | XP_016871668.1 | 4 | 245 | |||
XM_017016181.1 | XP_016871670.1 | 4 | 289 | |||
GenBank mRNA | AJ536328.2 | - | 4 | 464 | ||
AK122716.1 | - | 4 | 535 | |||
AK127756.1 | - | 4 | 371 | |||
AY572972.1 | - | 4 | 302 | |||
BC143380.1 | - | 4 | 242 |