Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D10C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256508.1 | NP_001243437.1 | 9 | 1198 | ||
NM_198517.3 | NP_940919.1 | 10 | 1409 | |||
NR_046266.1 | 9 | 1625 | ||||
XM_006718538.3 | XP_006718601.1 | 10 | 1668 | |||
XM_006718539.3 | XP_006718602.1 | 10 | 1651 | |||
XM_006718540.3 | XP_006718603.1 | 10 | 1581 | |||
XM_006718541.3 | XP_006718604.1 | 10 | 1672 | |||
XM_006718542.3 | XP_006718605.1 | 10 | 1655 | |||
XM_006718543.2 | XP_006718606.1 | 10 | 1389 | |||
XM_006718544.3 | XP_006718607.1 | 10 | 1583 | |||
XM_011545001.2 | XP_011543303.1 | 9 | 1610 | |||
GenBank mRNA | AK122585.1 | - | 3 | 4157 | ||
BC022202.1 | - | 9 | 1353 | |||
BC036873.1 | - | 9 | 1364 | 5224063 | ||
BC062999.1 | - | 10 | 1381 | 5176854 |