Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMIGO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198722.2 | NP_942015.1 | 1 | 633 | ||
GenBank mRNA | AY237003.1 | - | 1 | 293 | ||
AY358134.1 | - | 1 | 518 | |||
BC101363.1 | - | 1 | 454 | 40025738 | ||
BC101364.1 | - | 1 | 454 | 40025739 | ||
BC101365.2 | - | 1 | 454 | 40025741 | ||
BC110418.1 | - | 1 | 523 | 5211193 | ||
BQ673901.1 | - | 1 | 633 |