Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003032.2 | NP_003023.1 | 5 | 1072 | ||
NM_173216.2 | NP_775323.1 | 6 | 1395 | |||
NM_173217.2 | NP_775324.1 | 5 | 738 | |||
XM_005247719.1 | XP_005247776.1 | 5 | 983 | |||
XM_005247720.1 | XP_005247777.1 | 5 | 1041 | |||
XM_006713734.1 | XP_006713797.1 | 7 | 1650 | |||
XM_011513085.1 | XP_011511387.1 | 7 | 1658 | |||
XM_011513086.1 | XP_011511388.1 | 5 | 1041 | |||
XM_017007065.1 | XP_016862554.1 | 8 | 1742 | |||
XM_017007066.1 | XP_016862555.1 | 5 | 1070 | |||
XM_017007067.1 | XP_016862556.1 | 5 | 1190 | |||
XM_017007068.1 | XP_016862557.1 | 5 | 1075 | |||
GenBank mRNA | A17362.1 | - | 6 | 1199 | ||
A23699.1 | - | 3 | 776 | |||
AK128726.1 | - | 1 | 932 | |||
AK292879.1 | - | 5 | 1093 | |||
AK312023.1 | - | 2 | 101 | |||
BC031476.1 | - | 5 | 1066 | 4339124 | ||
BC040009.1 | - | 5 | 1072 | 5557868 | ||
DB235666.1 | - | 5 | 527 | |||
X17247.1 | - | 6 | 1193 | |||
X54363.1 | - | 5 | 1031 | |||
X62822.1 | - | 5 | 1072 |