Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRINP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199051.2 | NP_950252.1 | 3 | 845 | ||
XM_011509472.2 | XP_011507774.1 | 4 | 847 | |||
XM_011509475.2 | XP_011507777.1 | 3 | 585 | |||
XM_017001125.1 | XP_016856614.1 | 5 | 956 | |||
XM_017001126.1 | XP_016856615.1 | 4 | 821 | |||
XM_017001127.1 | XP_016856616.1 | 4 | 985 | |||
XM_017001128.1 | XP_016856617.1 | 4 | 2073 | |||
XM_017001131.1 | XP_016856620.1 | 4 | 935 | |||
XM_017001132.1 | XP_016856621.1 | 3 | 865 | |||
XM_017001133.1 | XP_016856622.1 | 3 | 865 | |||
GenBank mRNA | AB111893.1 | - | 3 | 592 | ||
AK123024.1 | - | 3 | 535 | |||
BC105052.1 | - | 2 | 416 | 8144055 | ||
BC105054.1 | - | 2 | 416 | 8144057 |