Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFT2D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199344.2 | NP_955376.1 | 4 | 344 | ||
GenBank mRNA | AK290325.1 | - | 4 | 344 | ||
AL035297.1 | - | 4 | 353 | |||
AY358675.1 | - | 2 | 62 | |||
BC068098.1 | - | 4 | 340 | 6304345 |