Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290049.1 | NP_001276978.1 | 26 | 3210 | ||
NM_003715.3 | NP_003706.2 | 24 | 3177 | |||
XM_006714396.3 | XP_006714459.1 | 25 | 3193 | |||
XM_006714397.3 | XP_006714460.1 | 25 | 3189 | |||
GenBank mRNA | AK314289.1 | - | 24 | 3067 | ||
AL832010.2 | - | 26 | 3133 | |||
AL833753.1 | - | 15 | 1909 | |||
BC032654.1 | - | 24 | 3032 | 5533256 | ||
D86326.1 | - | 24 | 3054 |