Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003943.4 | NP_003934.1 | 2 | 2181 | ||
GenBank mRNA | AF062534.1 | - | 2 | 1564 | ||
AK002155.1 | - | 1 | 1085 | |||
AK123795.1 | - | 2 | 2181 | |||
BC022301.1 | - | 2 | 1507 | 4694574 |
Gene Symbol: FAM47E-STBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242939.1 | NP_001229868.1 | 7 | 2290 | ||
GenBank mRNA | AK090914.1 | - | 5 | 1763 | ||
AK092277.1 | - | 7 | 2269 | |||
AK316039.1 | - | 3 | 1446 |