Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIAO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004804.2 | NP_004795.1 | 5 | 756 | ||
GenBank mRNA | AK223257.1 | - | 5 | 677 | ||
AK299503.1 | - | 5 | 443 | |||
AK315568.1 | - | 5 | 665 | |||
BC001395.2 | - | 5 | 620 | 3050587 | ||
BC032812.1 | - | 5 | 650 | 5219740 | ||
CR456802.1 | - | 5 | 551 | |||
EF011618.1 | - | 5 | 650 | |||
U63810.1 | - | 5 | 707 |