Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135585.1 | NP_001129057.1 | 4, 5 | 718 | ||
NM_003039.2 | NP_003030.1 | 4, 5 | 718 | |||
XM_005263491.3 | XP_005263548.1 | 5, 6 | 516 | |||
XM_017002133.1 | XP_016857622.1 | 6, 7 | 744 | |||
XM_017002134.1 | XP_016857623.1 | 6, 7 | 710 | |||
XM_017002135.1 | XP_016857624.1 | 6, 7 | 804 | |||
XM_017002136.1 | XP_016857625.1 | 6, 7 | 780 | |||
XM_017002137.1 | XP_016857626.1 | 7, 8 | 816 | |||
XM_017002138.1 | XP_016857627.1 | 7, 8 | 792 | |||
XM_017002139.1 | XP_016857628.1 | 5, 6 | 726 | |||
XM_017002140.1 | XP_016857629.1 | 6, 7 | 647 | |||
XM_017002141.1 | XP_016857630.1 | 5, 6 | 549 | |||
XM_017002142.1 | XP_016857631.1 | 2, 3 | 289 | |||
GenBank mRNA | AF451839.1 | - | 2, 3 | 231 | ||
AF479408.1 | - | 2, 3 | 231 | |||
AK289849.1 | - | 4, 5 | 474 | |||
AK290398.1 | - | 4, 5 | 508 | |||
AK294368.1 | - | 4, 5 | 504 | |||
AK295785.1 | - | 3, 4 | 361 | |||
AK300091.1 | - | 2, 3 | 175 | |||
AK300477.1 | - | 2, 3 | 294 | |||
AK302047.1 | - | 4, 5 | 359 | |||
AK302057.1 | - | 7, 8 | 642 | |||
AK302137.1 | - | 6, 7 | 594 | |||
BC001692.2 | - | 4, 5 | 477 | 2966459 | ||
BC001820.2 | - | 4, 5 | 477 | 2966459 | ||
BC035878.1 | - | 4, 5 | 505 | |||
M55531.1 | - | 4, 5 | 501 |