Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIAO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004804.2 | NP_004795.1 | 7 | 1234 | ||
GenBank mRNA | AK056423.1 | - | 5 | 1600 | ||
AK124703.1 | - | 1 | 837 | |||
AK223257.1 | - | 7 | 1155 | |||
AK299503.1 | - | 7 | 921 | |||
BC001395.2 | - | 7 | 1098 | 3050587 | ||
BC032812.1 | - | 7 | 1128 | 5219740 | ||
EF011618.1 | - | 7 | 1128 |