Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010938.1 | NP_001010938.1 | 13 | 2954 | ||
NM_001308046.1 | NP_001294975.1 | 12 | 2967 | |||
NM_005781.4 | NP_005772.3 | 12 | 3118 | |||
XM_005269268.3 | XP_005269325.1 | 13 | 2954 | |||
XM_005269270.3 | XP_005269327.1 | 13 | 2802 | |||
XM_005269274.3 | XP_005269331.1 | 10 | 2219 | |||
XM_005269275.3 | XP_005269332.1 | 12 | 2698 | |||
XM_011512317.2 | XP_011510619.1 | 15 | 3521 | |||
XM_011512318.2 | XP_011510620.2 | 13 | 2893 | |||
XM_011512320.1 | XP_011510622.1 | 13 | 2676 | |||
XM_011512321.2 | XP_011510623.1 | 11 | 2493 | |||
XM_017005508.1 | XP_016860997.1 | 12 | 2848 | |||
XM_017005509.1 | XP_016860998.1 | 12 | 2848 | |||
XM_017005510.1 | XP_016860999.1 | 13 | 2893 | |||
GenBank mRNA | AK126712.1 | - | 1 | 2015 | ||
AK127455.1 | - | 4 | 1276 | |||
AK131539.1 | - | 13 | 2954 | |||
AK225786.1 | - | 12 | 2914 | |||
BE676668.1 | - | 2 | 458 | |||
L13738.2 | - | 12 | 3187 |