Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005879.2 | NP_005870.2 | 13, 14 | 1340 | ||
XM_011533264.1 | XP_011531566.1 | 13, 14 | 1434 | |||
XM_017005526.1 | XP_016861015.1 | 10, 11 | 1068 | |||
GenBank mRNA | AB451320.1 | - | 13, 14 | 1234 | ||
AB451449.1 | - | 13, 14 | 1234 | |||
AK225873.1 | - | 10, 11 | 1019 | |||
AK292172.1 | - | 13, 14 | 1328 | |||
BC000310.2 | - | 13, 14 | 1321 | 2821007 | ||
BC019283.1 | - | 13, 14 | 1321 | 2821007 | ||
U77845.1 | - | 13, 14 | 1336 |