Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005879.2 | NP_005870.2 | 15 | 1445 | ||
XM_011533264.1 | XP_011531566.1 | 15 | 1539 | |||
XM_017005526.1 | XP_016861015.1 | 12 | 1173 | |||
GenBank mRNA | AB451320.1 | - | 15 | 1339 | ||
AB451449.1 | - | 15 | 1339 | |||
AK225873.1 | - | 12 | 1124 | |||
AK292172.1 | - | 15 | 1433 | |||
BC000310.2 | - | 15 | 1426 | 2821007 | ||
BC019283.1 | - | 15 | 1426 | 2821007 | ||
U77845.1 | - | 15 | 1441 |