Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL6IP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006407.3 | NP_006398.1 | 3 | 1641 | ||
GenBank mRNA | AB097051.1 | - | 3 | 1621 | ||
AF070523.1 | - | 3 | 1648 | |||
AF161476.1 | - | 3 | 1621 | |||
AK074612.1 | - | 3 | 1594 | |||
BC020797.1 | - | 3 | 1559 | 4134658 | ||
BC107899.1 | - | 3 | 989 |