Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320044.1 | NP_001306973.1 | 10 | 1695 | ||
NM_012235.3 | NP_036367.2 | 13 | 2180 | |||
XM_005264967.1 | XP_005265024.1 | 13 | 2107 | |||
XM_005264968.1 | XP_005265025.1 | 10 | 1849 | |||
XM_005264971.1 | XP_005265028.1 | 7 | 1260 | |||
XM_011533501.1 | XP_011531803.1 | 13 | 2207 | |||
XM_011533502.2 | XP_011531804.1 | 5 | 856 | |||
XM_017005918.1 | XP_016861407.1 | 13 | 2194 | |||
XM_017005919.1 | XP_016861408.1 | 13 | 2127 | |||
XM_017005920.1 | XP_016861409.1 | 13 | 2127 | |||
XM_017005921.1 | XP_016861410.1 | 7 | 1260 | |||
GenBank mRNA | AK027402.1 | - | 7 | 1173 | ||
AK291484.1 | - | 8 | 1373 | |||
AK300311.1 | - | 10 | 1585 | |||
BC020987.1 | - | 5 | 901 | 3852258 | ||
D83782.2 | - | 13 | 2166 |